2,3-Dimethoxy-6-(methylthio)phenol ≥97%

Supplier: AOBChem USA

Warning

60095-500MG 60095-1G 60095-5G 60095-25G
77330-784EA 255.44 USD
77330-784 77354-950 77366-998 77388-588
2,3-Dimethoxy-6-(methylthio)phenol ≥97%
2,3-Dimethoxy-6-(methylthio)phenol

Precaución: for Research Use Only

Formula: C₉H₁₂O₃S
Storage Temperature: Ambient
MDL Number: MFCD34759597
CAS Number: 2711589-85-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR