3-Hydroxy-3-mesityl-N-methoxy-N-methylpropanamide ≥95%
Supplier: AOBChem USA
|
Warning
|
60082-1G
60082-5G
60082-25G
77341-170EA
72.1
USD
77341-170
77366-996
77379-056
3-Hydroxy-3-mesityl-N-methoxy-N-methylpropanamide ≥95%
3-Hydroxy-3-mesityl-N-methoxy-N-methylpropanamide
Precaución: for Research Use Only
Formula:
C₁₄H₂₁NO₃ MW: 251.321 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34759593 CAS Number: 2755718-69-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...