3-Hydroxy-N-methoxy-N-methyl-3-(4-pentylphenyl)propanamide ≥95%

Supplier: AOBChem USA

Warning

60080-500MG 60080-1G 60080-5G 60080-25G
77330-782EA 120.51 USD
77330-782 77343-766 77366-994 77379-054
3-Hydroxy-N-methoxy-N-methyl-3-(4-pentylphenyl)propanamide ≥95%
3-Hydroxy-N-methoxy-N-methyl-3-(4-pentylphenyl)propanamide

Precaución: for Research Use Only

Formula: C₁₆H₂₅NO₃
MW: 279.375 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759591
CAS Number: 2807471-04-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR