Ethyl 3-(3-bromo-5-(trifluoromethyl)phenyl)-3-hydroxypropanoate ≥97%

Supplier: AOBChem USA

Warning

60070-1G 60070-5G 60070-25G
77341-042EA 66.95 USD
77341-042 77366-992 77379-052
Ethyl 3-(3-bromo-5-(trifluoromethyl)phenyl)-3-hydroxypropanoate ≥97%
Ethyl 3-(3-bromo-5-(trifluoromethyl)phenyl)-3-hydroxypropanoate

Precaución: for Research Use Only

Formula: C₁₂H₁₂BrF₃O₃
Storage Temperature: Ambient
MDL Number: MFCD34759584
CAS Number: 2755718-48-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR