3-Hydroxy-N-methoxy-N-methyl-3-(2-(trifluoromethoxy)phenyl)propanamide ≥97%

Supplier: AOBChem USA

Warning

60067-500MG 60067-1G 60067-5G 60067-25G
77330-780EA 126.69 USD
77330-780 77344-170 77366-990 77379-050
3-Hydroxy-N-methoxy-N-methyl-3-(2-(trifluoromethoxy)phenyl)propanamide ≥97%
3-Hydroxy-N-methoxy-N-methyl-3-(2-(trifluoromethoxy)phenyl)propanamide

Precaución: for Research Use Only

Formula: C₁₂H₁₄F₃NO₄
MW: 293.239 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759581
CAS Number: 2755719-50-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR