Ethyl 3-hydroxy-3-(2-(trifluoromethoxy)phenyl)propanoate ≥95%

Supplier: AOBChem USA

Warning

60066-1G 60066-5G 60066-25G
77340-762EA 60.77 USD
77340-762 77366-988 77379-048
Ethyl 3-hydroxy-3-(2-(trifluoromethoxy)phenyl)propanoate ≥95%
Ethyl 3-hydroxy-3-(2-(trifluoromethoxy)phenyl)propanoate

Precaución: for Research Use Only

Formula: C₁₂H₁₃F₃O₄
MW: 278.224 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17338861
CAS Number: 1271063-49-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR