1-Bromo-4-isopropoxy-5-methoxy-2-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

60016-500MG 60016-1G 60016-5G 60016-25G
77330-774EA 183.34 USD
77330-774 77347-124 77366-982 77388-582
1-Bromo-4-isopropoxy-5-methoxy-2-(trifluoromethyl)benzene ≥97%
1-Bromo-4-isopropoxy-5-methoxy-2-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₁₁H₁₂BrF₃O₂
MW: 313.111 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759553
CAS Number: 2807447-96-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR