1-(Benzyloxy)-4-bromo-2-methoxy-5-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

60015-500MG 60015-1G 60015-5G 60015-25G
77330-772EA 202.91 USD
77330-772 77354-942 77366-980 77388-580
1-(Benzyloxy)-4-bromo-2-methoxy-5-(trifluoromethyl)benzene ≥97%
1-(Benzyloxy)-4-bromo-2-methoxy-5-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₁₅H₁₂BrF₃O₂
MW: 361.154 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759552
CAS Number: 2807455-57-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR