1-Bromo-4-ethoxy-5-methoxy-2-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

60014-500MG 60014-1G 60014-5G 60014-25G
77330-770EA 260.59 USD
77330-770 77354-940 77366-978 77388-578
1-Bromo-4-ethoxy-5-methoxy-2-(trifluoromethyl)benzene ≥97%
1-Bromo-4-ethoxy-5-methoxy-2-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₁₀H₁₀BrF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD34759551
CAS Number: 2384667-82-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR