Ethyl 4-fluoro-3-formyl-5-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

60011-500MG 60011-1G 60011-5G
77330-766EA 329.6 USD
77330-766 77354-938 77379-042
Ethyl 4-fluoro-3-formyl-5-methylbenzoate ≥97%
Ethyl 4-fluoro-3-formyl-5-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₁FO₃
MW: 210.202 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759548
CAS Number: 2387270-10-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR