(2,4-Dichloro-3-iodophenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

60009-500MG 60009-1G 60009-5G
77330-764EA 326.51 USD
77330-764 77354-936 77379-040
(2,4-Dichloro-3-iodophenyl)methanol ≥97%
(2,4-Dichloro-3-iodophenyl)methanol

Precaución: for Research Use Only

Formula: C₇H₅Cl₂IO
MW: 302.924 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759546
CAS Number: 2095235-20-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR