Methyl 4-ethoxy-2-(trifluoromethyl)benzoate ≥97%

Supplier: AOBChem USA

Warning

59983-500MG 59983-1G 59983-5G
77330-760EA 334.75 USD
77330-760 77354-932 77379-036
Methyl 4-ethoxy-2-(trifluoromethyl)benzoate ≥97%
Methyl 4-ethoxy-2-(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₁F₃O₃
Storage Temperature: Ambient
MDL Number: MFCD34759534
CAS Number: 2764730-96-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR