(3-Bromo-4-butoxyphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

59981-500MG 59981-1G 59981-5G 59981-25G
77330-758EA 168.92 USD
77330-758 77346-290 77366-974 77388-574
(3-Bromo-4-butoxyphenyl)methanol ≥97%
(3-Bromo-4-butoxyphenyl)methanol

Precaución: for Research Use Only

Formula: C₁₁H₁₅BrO₂
MW: 259.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17284400
CAS Number: 1343138-34-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR