Methyl 3-(3-bromo-4-methoxyphenyl)-3-hydroxypropanoate ≥95%

Supplier: AOBChem USA

Warning

59980-500MG 59980-1G 59980-5G 59980-25G
77330-756EA 147.29 USD
77330-756 77345-226 77366-972 77388-572
Methyl 3-(3-bromo-4-methoxyphenyl)-3-hydroxypropanoate ≥95%
Methyl 3-(3-bromo-4-methoxyphenyl)-3-hydroxypropanoate

Precaución: for Research Use Only

Formula: C₁₁H₁₃BrO₄
MW: 289.123 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759533
CAS Number: 2228787-35-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR