3-(4-Bromo-2-methylphenyl)-3-hydroxy-N-methoxy-N-methylpropanamide ≥97%
Supplier: AOBChem USA
|
Warning
|
59969-500MG
59969-1G
59969-5G
59969-25G
77330-752EA
179.22
USD
77330-752
77346-732
77366-966
77388-568
3-(4-Bromo-2-methylphenyl)-3-hydroxy-N-methoxy-N-methylpropanamide ≥97%
3-(4-Bromo-2-methylphenyl)-3-hydroxy-N-methoxy-N-methylpropanamide
Precaución: for Research Use Only
Formula:
C₁₂H₁₆BrNO₃ MW: 302.164 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34759525 CAS Number: 2755723-79-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...