3-(3-Ethyl-4-methoxyphenyl)-3-hydroxy-N-methoxy-N-methylpropanamide ≥97%

Supplier: AOBChem USA

Warning

59964-500MG 59964-1G 59964-5G
77338-780EA 517.06 USD
77338-780 77354-930 77379-032
3-(3-Ethyl-4-methoxyphenyl)-3-hydroxy-N-methoxy-N-methylpropanamide ≥97%
3-(3-Ethyl-4-methoxyphenyl)-3-hydroxy-N-methoxy-N-methylpropanamide

Precaución: for Research Use Only

Formula: C₁₄H₂₁NO₄
MW: 267.321 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759523
CAS Number: 2807473-92-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR