Methyl 3-(methylthio)-4-(trifluoromethoxy)benzoate ≥95%

Supplier: AOBChem USA

Warning

59952-500MG 59952-1G 59952-5G
77339-444EA 590.19 USD
77339-444 77354-926 77379-028
Methyl 3-(methylthio)-4-(trifluoromethoxy)benzoate ≥95%
Methyl 3-(methylthio)-4-(trifluoromethoxy)benzoate

Precaución: for Research Use Only

Formula: C₁₀H₉F₃O₃S
MW: 266.237 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759511
CAS Number: 2901098-29-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR