(5-Bromo-2-(trifluoromethoxy)phenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

59948-500MG 59948-1G 59948-5G
77338-222EA 469.68 USD
77338-222 77354-922 77379-024
(5-Bromo-2-(trifluoromethoxy)phenyl)(methyl)sulfane ≥97%
(5-Bromo-2-(trifluoromethoxy)phenyl)(methyl)sulphane

Precaución: for Research Use Only

Formula: C₈H₆BrF₃OS
Storage Temperature: Ambient
MDL Number: MFCD34475366
CAS Number: 2169014-04-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR