3-Hydroxy-4-(trifluoromethoxy)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

59945-500MG 59945-1G 59945-5G 59945-25G
77330-748EA 175.1 USD
77330-748 77346-490 77366-964 77388-564
3-Hydroxy-4-(trifluoromethoxy)benzaldehyde ≥95%
3-Hydroxy-4-(trifluoromethoxy)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₅F₃O₃
MW: 206.119 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13250180
CAS Number: 1208078-41-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR