(3-(Benzyloxy)-4-(trifluoromethoxy)phenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

59936-500MG 59936-1G 59936-5G
77338-994EA 541.78 USD
77338-994 77354-920 77379-022
(3-(Benzyloxy)-4-(trifluoromethoxy)phenyl)methanol ≥97%
(3-(Benzyloxy)-4-(trifluoromethoxy)phenyl)methanol

Precaución: for Research Use Only

Formula: C₁₅H₁₃F₃O₃
MW: 298.257 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759503
CAS Number: 2764729-02-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR