Methyl 3-(benzyloxy)-4-(trifluoromethoxy)benzoate ≥97%

Supplier: AOBChem USA

Warning

59934-500MG 59934-1G 59934-5G
77339-692EA 614.91 USD
77339-692 77354-918 77379-020
Methyl 3-(benzyloxy)-4-(trifluoromethoxy)benzoate ≥97%
Methyl 3-(benzyloxy)-4-(trifluoromethoxy)benzoate

Precaución: for Research Use Only

Formula: C₁₆H₁₃F₃O₄
Storage Temperature: Ambient
MDL Number: MFCD34759501
CAS Number: 2764730-92-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR