3-(Benzyloxy)-4-(trifluoromethoxy)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

59933-500MG 59933-1G 59933-5G
77337-774EA 441.87 USD
77337-774 77354-916 77379-018
3-(Benzyloxy)-4-(trifluoromethoxy)benzoic acid ≥97%
3-(Benzyloxy)-4-(trifluoromethoxy)benzoic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₁F₃O₄
MW: 312.241 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759500
CAS Number: 2755721-29-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR