3-(Difluoromethoxy)-4-(trifluoromethoxy)benzoic acid ≥95%

Supplier: AOBChem USA

Warning

59925-500MG 59925-1G 59925-5G
77338-638EA 505.73 USD
77338-638 77354-910 77379-012
3-(Difluoromethoxy)-4-(trifluoromethoxy)benzoic acid ≥95%
3-(Difluoromethoxy)-4-(trifluoromethoxy)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₅F₅O₄
MW: 272.126 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759493
CAS Number: 2764731-12-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR