4-Bromo-1-methyl-5-(methylthio)-1H-pyrazole ≥97%

Supplier: AOBChem USA

Warning

59909-500MG 59909-1G 59909-5G
77337-674EA 435.69 USD
77337-674 77354-904 77379-008
4-Bromo-1-methyl-5-(methylthio)-1H-pyrazole ≥97%
4-Bromo-1-methyl-5-(methylthio)-1H-pyrazole

Precaución: for Research Use Only

Formula: C₅H₇BrN₂S
MW: 207.091 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32678837
CAS Number: 2418642-17-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR