4-Bromo-2-(difluoromethoxy)-1-(trifluoromethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

59907-500MG 59907-1G 59907-5G
77337-462EA 414.06 USD
77337-462 77354-902 77379-006
4-Bromo-2-(difluoromethoxy)-1-(trifluoromethoxy)benzene ≥97%
4-Bromo-2-(difluoromethoxy)-1-(trifluoromethoxy)benzene

Precaución: for Research Use Only

Formula: C₈H₄BrF₅O₂
MW: 307.012 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28738162
CAS Number: 1807244-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR