2-(Benzyloxy)-4-bromo-1-(trifluoromethoxy)benzene ≥95%

Supplier: AOBChem USA

Warning

59905-500MG 59905-1G 59905-25G 59905-5G
77330-742EA 292.52 USD
77330-742 77354-900 77388-558 77379-004
2-(Benzyloxy)-4-bromo-1-(trifluoromethoxy)benzene ≥95%
2-(Benzyloxy)-4-bromo-1-(trifluoromethoxy)benzene

Precaución: for Research Use Only

Formula: C₁₄H₁₀BrF₃O₂
MW: 347.127 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759476
CAS Number: 2241577-05-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR