(3-Bromo-4-propoxyphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

59897-500MG 59897-1G 59897-5G 59897-25G
77330-738EA 123.6 USD
77330-738 77344-060 77366-956 77378-998
(3-Bromo-4-propoxyphenyl)methanol ≥97%
(3-Bromo-4-propoxyphenyl)methanol

Precaución: for Research Use Only

Formula: C₁₀H₁₃BrO₂
MW: 245.113 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08691776
CAS Number: 915922-40-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR