2-(Benzyloxy)-4-iodo-1-(trifluoromethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

59896-500MG 59896-1G 59896-5G
77339-806EA 647.87 USD
77339-806 77354-898 77378-996
2-(Benzyloxy)-4-iodo-1-(trifluoromethoxy)benzene ≥97%
2-(Benzyloxy)-4-iodo-1-(trifluoromethoxy)benzene

Precaución: for Research Use Only

Formula: C₁₄H₁₀F₃IO₂
MW: 394.128 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759472
CAS Number: 2241577-06-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR