Ethyl 2-bromo-4-fluoro-5-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

59892-500MG 59892-1G 59892-5G
77338-778EA 517.06 USD
77338-778 77354-896 77378-992
Ethyl 2-bromo-4-fluoro-5-methoxybenzoate ≥97%
Ethyl 2-bromo-4-fluoro-5-methoxybenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₀BrFO₃
MW: 277.087 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759469
CAS Number: 2168488-75-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR