2-Fluoro-1-(methoxymethyl)-4-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

59890-1G 59890-5G 59890-25G
77341-808EA 104.03 USD
77341-808 77366-952 77378-990
2-Fluoro-1-(methoxymethyl)-4-methylbenzene ≥97%
2-Fluoro-1-(methoxymethyl)-4-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₁FO
MW: 154.181 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22491720
CAS Number: 1824065-67-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR