(3-Fluoro-4-(trifluoromethoxy)phenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

59883-500MG 59883-1G 59883-5G 59883-25G
77330-734EA 146.26 USD
77330-734 77345-174 77366-950 77388-556
(3-Fluoro-4-(trifluoromethoxy)phenyl)(methyl)sulfane ≥95%
(3-Fluoro-4-(trifluoromethoxy)phenyl)(methyl)sulphane

Precaución: for Research Use Only

Formula: C₈H₆F₄OS
MW: 226.191 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23143145
CAS Number: 1443312-18-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR