Ethyl 5-bromo-2-methoxyisonicotinate ≥95%

Supplier: AOBChem USA

Warning

59879-500MG 59879-1G 59879-5G 59879-25G
77330-732EA 270.89 USD
77330-732 77354-890 77366-948 77388-554
Ethyl 5-bromo-2-methoxyisonicotinate ≥95%
Ethyl 5-bromo-2-methoxyisonicotinate

Precaución: for Research Use Only

Formula: C₉H₁₀BrNO₃
Storage Temperature: Ambient
MDL Number: MFCD14698345
CAS Number: 1214357-96-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR