Benzyl 2-fluorobenzoate ≥95%

Supplier: AOBChem USA

Warning

59872-500MG 59872-1G 59872-5G 59872-25G
77330-730EA 204.97 USD
77330-730 77354-886 77366-946 77388-552
Benzyl 2-fluorobenzoate ≥95%
Benzyl 2-fluorobenzoate

Precaución: for Research Use Only

Formula: C₁₄H₁₁FO₂
MW: 230.234 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04341248
CAS Number: 478059-14-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR