Ethyl 3-chloro-4-phenoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

59871-500MG 59871-1G 59871-5G 59871-25G
77330-728EA 258.53 USD
77330-728 77354-884 77366-944 77388-550
Ethyl 3-chloro-4-phenoxybenzoate ≥97%
Ethyl 3-chloro-4-phenoxybenzoate

Precaución: for Research Use Only

Formula: C₁₅H₁₃ClO₃
Storage Temperature: Ambient
MDL Number: MFCD34759458
CAS Number: 2755721-24-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR