Methyl 4-methyl-6-(methylthio)nicotinate ≥97%

Supplier: AOBChem USA

Warning

59867-500MG 59867-1G 59867-5G
77336-838EA 394.49 USD
77336-838 77354-880 77378-982
Methyl 4-methyl-6-(methylthio)nicotinate ≥97%
Methyl 4-methyl-6-(methylthio)nicotinate

Precaución: for Research Use Only

Formula: C₉H₁₁NO₂S
Storage Temperature: Ambient
MDL Number: MFCD21088016
CAS Number: 1355207-49-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR