(3-Bromo-5-fluoro-2-methoxyphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

59852-500MG 59852-1G 59852-5G 59852-25G
77330-718EA 105.06 USD
77330-718 77343-094 77366-934 77378-974
(3-Bromo-5-fluoro-2-methoxyphenyl)(methyl)sulfane ≥97%
(3-Bromo-5-fluoro-2-methoxyphenyl)(methyl)sulphane

Precaución: for Research Use Only

Formula: C₈H₈BrFOS
Storage Temperature: Ambient
MDL Number: MFCD34759449
CAS Number: 2091914-14-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR