(3-Bromo-5-iodo-4-methoxyphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

59849-500MG 59849-1G 59849-5G
77337-304EA 410.97 USD
77337-304 77354-870 77378-972
(3-Bromo-5-iodo-4-methoxyphenyl)methanol ≥97%
(3-Bromo-5-iodo-4-methoxyphenyl)methanol

Precaución: for Research Use Only

Formula: C₈H₈BrIO₂
MW: 342.956 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759447
CAS Number: 2383341-29-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR