3-Bromo-4-methoxy-5-(methylthio)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

59846-500MG 59846-1G 59846-5G
77338-776EA 517.06 USD
77338-776 77354-866 77378-968
3-Bromo-4-methoxy-5-(methylthio)benzaldehyde ≥97%
3-Bromo-4-methoxy-5-(methylthio)benzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉BrO₂S
Storage Temperature: Ambient
MDL Number: MFCD34759445
CAS Number: 71125-91-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR