1-Ethoxy-2-fluoro-3-iodo-5-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

59837-500MG 59837-1G 59837-5G 59837-25G
77330-714EA 152.44 USD
77330-714 77345-462 77366-930 77388-540
1-Ethoxy-2-fluoro-3-iodo-5-methylbenzene ≥97%
1-Ethoxy-2-fluoro-3-iodo-5-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₀FIO
Storage Temperature: Ambient
MDL Number: MFCD34759441
CAS Number: 2387304-26-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR