(3-Ethoxy-2-fluoro-5-methylphenyl)(methyl)sulphane ≥97%

Supplier: AOBChem USA

Warning

59836-500MG 59836-1G 59836-5G 59836-25G
77330-712EA 147.29 USD
77330-712 77345-224 77366-928 77388-538
(3-Ethoxy-2-fluoro-5-methylphenyl)(methyl)sulphane ≥97%
(3-Ethoxy-2-fluoro-5-methylphenyl)(methyl)sulphane

Precaución: for Research Use Only

Formula: C₁₀H₁₃FOS
Storage Temperature: Ambient
MDL Number: MFCD34759440
CAS Number: 2706074-27-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR