Ethyl 3-ethoxy-2-fluoro-5-methylbenzoate ≥95%

Supplier: AOBChem USA

Warning

59833-500MG 59833-1G 59833-5G 59833-25G
77330-710EA 241.02 USD
77330-710 77354-862 77366-926 77388-536
Ethyl 3-ethoxy-2-fluoro-5-methylbenzoate ≥95%
Ethyl 3-ethoxy-2-fluoro-5-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₅FO₃
MW: 226.244 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759437
CAS Number: 2384761-18-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR