2-(Benzyloxy)-1-fluoro-4-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

59811-500MG 59811-1G 59811-5G 59811-25G
77330-700EA 118.45 USD
77330-700 77343-566 77366-914 77378-960
2-(Benzyloxy)-1-fluoro-4-methylbenzene ≥97%
2-(Benzyloxy)-1-fluoro-4-methylbenzene

Precaución: for Research Use Only

Formula: C₁₄H₁₃FO
MW: 216.251 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12405675
CAS Number: 1260881-29-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR