5-Chloro-2-isopropylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

59806-500MG 59806-1G 59806-5G 59806-25G
77330-698EA 239.99 USD
77330-698 77354-856 77366-912 77388-528
5-Chloro-2-isopropylbenzoic acid ≥97%
5-Chloro-2-isopropylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClO₂
MW: 198.646 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28356768
CAS Number: 1369813-02-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR