3-Chloro-2-formyl-6-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

59804-500MG 59804-1G 59804-5G
77337-866EA 449.08 USD
77337-866 77354-854 77378-958
3-Chloro-2-formyl-6-methylbenzoic acid ≥97%
3-Chloro-2-formyl-6-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₉H₇ClO₃
MW: 198.603 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759421
CAS Number: 2384766-48-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR