3-Chloro-6-methyl-2-(methylthio)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

59795-500MG 59795-1G 59795-5G
77330-694EA 329.6 USD
77330-694 77354-850 77378-956
3-Chloro-6-methyl-2-(methylthio)benzaldehyde ≥97%
3-Chloro-6-methyl-2-(methylthio)benzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉ClOS
Storage Temperature: Ambient
MDL Number: MFCD34759412
CAS Number: 2901086-24-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR