4-Bromo-2-fluoro-3-methoxyphenol ≥97%

Supplier: AOBChem USA

Warning

59793-500MG 59793-1G 59793-5G 59793-25G
77330-692EA 231.75 USD
77330-692 77354-848 77366-908 77388-524
4-Bromo-2-fluoro-3-methoxyphenol ≥97%
4-Bromo-2-fluoro-3-methoxyphenol

Precaución: for Research Use Only

Formula: C₇H₆BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD28738850
CAS Number: 1779753-99-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR