1-Fluoro-5-iodo-2,3-dimethylbenzene ≥97%

Supplier: AOBChem USA

Warning

59792-500MG 59792-1G 59792-5G
77340-032EA 718.94 USD
77340-032 77354-846 77378-954
1-Fluoro-5-iodo-2,3-dimethylbenzene ≥97%
1-Fluoro-5-iodo-2,3-dimethylbenzene

Precaución: for Research Use Only

Formula: C₈H₈FI
MW: 250.052 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28794425
CAS Number: 1803852-65-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR