2-Chloro-4-iodo-3-(methylthio)pyridine ≥95%
Supplier: AOBChem USA
|
Warning
|
59785-500MG
59785-1G
59785-5G
59785-25G
77330-690EA
220.42
USD
77330-690
77354-844
77366-906
77388-522
2-Chloro-4-iodo-3-(methylthio)pyridine ≥95%
2-Chloro-4-iodo-3-(methylthio)pyridine
Precaución: for Research Use Only
Formula:
C₆H₅ClINS MW: 285.533 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34759407 CAS Number: 2764729-17-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...