(3-Iodo-2-methoxyphenyl)(methyl)sulphane ≥97%

Supplier: AOBChem USA

Warning

59775-500MG 59775-1G 59775-5G 59775-25G
77330-684EA 204.97 USD
77330-684 77354-838 77366-900 77388-516
(3-Iodo-2-methoxyphenyl)(methyl)sulphane ≥97%
(3-Iodo-2-methoxyphenyl)(methyl)sulphane

Precaución: for Research Use Only

Formula: C₈H₉IOS
Storage Temperature: Ambient
MDL Number: MFCD22489428
CAS Number: 1822764-26-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR