2-Ethoxy-3-(methylthio)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

59766-500MG 59766-1G 59766-25G 59766-5G
77330-680EA 296.64 USD
77330-680 77354-834 77388-512 77378-952
2-Ethoxy-3-(methylthio)benzoic acid ≥97%
2-Ethoxy-3-(methylthio)benzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₃S
Storage Temperature: Ambient
MDL Number: MFCD34759393
CAS Number: 2755723-29-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR